siRNA Details
Gene Symbol: Sept4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011893.1 | NP_001011893.1 | 5 | 511 | ||
XM_008767966.1 | XP_008766188.1 | 9 | 1691 | |||
GenBank mRNA | AY208293.1 | - | 5 | 511 | ||
AY208294.1 | - | 5 | 511 | |||
BC088334.1 | - | 4 | 359 | |||
BC126069.1 | - | 5 | 412 | 7441949 |
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053931.3 | NP_446383.3 | 5 | 687 | ||
GenBank mRNA | AB027143.1 | - | 5 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
