siRNA Details
Gene Symbol: Mcf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053951.2 | NP_446403.2 | 8 | 853 | ||
XM_006253416.2 | XP_006253478.1 | 8 | 4115 | |||
XM_006253419.2 | XP_006253481.1 | 8 | 4115 | |||
XM_006253421.2 | XP_006253483.1 | 8 | 4114 | |||
XM_006253423.2 | XP_006253485.1 | 8 | 2741 | |||
XM_006253424.2 | XP_006253486.1 | 8 | 2932 | |||
XM_008771381.1 | XP_008769603.1 | 10 | 2083 | |||
XM_008771382.1 | XP_008769604.1 | 8 | 1485 | |||
XM_008771383.1 | XP_008769605.1 | 8 | 1768 | |||
GenBank mRNA | Z35654.1 | - | 6 | 949 |
