siRNA Details
Gene Symbol: Mcf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053951.2 | NP_446403.2 | 9, 10 | 1022 | ||
XM_006253416.2 | XP_006253478.1 | 9, 10 | 4284 | |||
XM_006253419.2 | XP_006253481.1 | 9, 10 | 4284 | |||
XM_006253421.2 | XP_006253483.1 | 9, 10 | 4283 | |||
XM_006253423.2 | XP_006253485.1 | 9, 10 | 2910 | |||
XM_006253424.2 | XP_006253486.1 | 9, 10 | 3101 | |||
XM_008771381.1 | XP_008769603.1 | 11, 12 | 2252 | |||
XM_008771382.1 | XP_008769604.1 | 9, 10 | 1654 | |||
XM_008771383.1 | XP_008769605.1 | 9, 10 | 1937 | |||
GenBank mRNA | Z35654.1 | - | 7, 8 | 1118 |
