siRNA Details
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057132.3 | NP_476473.1 | 4 | 549 | ||
XM_006243699.1 | XP_006243761.1 | 6 | 817 | |||
XM_006243700.1 | XP_006243762.1 | 6 | 792 | |||
XM_006243701.1 | XP_006243763.1 | 6 | 912 | |||
GenBank mRNA | AY026068.1 | - | 4 | 549 | ||
AY026069.1 | - | 4 | 549 | |||
BC061732.1 | - | 6 | 726 | 5598621 |
