siRNA Details
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057142.1 | NP_476483.1 | 3, 4 | 397 | ||
GenBank mRNA | EU348640.1 | - | 2, 3 | 320 | ||
EU348642.1 | - | 3, 4 | 397 | |||
EU348643.1 | - | 3, 4 | 397 | |||
EU348644.1 | - | 3, 4 | 397 | |||
U66707.1 | - | 3, 4 | 397 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081358, AK122498, AK141618, BC138450.
Gene Ontology Categories:
