siRNA Details
Gene Symbol: Manea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080785.2 | NP_542963.2 | 1 | 183 | ||
XM_006237960.1 | XP_006238022.1 | 2 | 487 | |||
XM_006237961.2 | XP_006238023.1 | 2 | 406 | |||
GenBank mRNA | AY599499.1 | - | 1 | 183 | ||
FQ223109.1 | - | 2 | 412 |
