siRNA Details
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007238.4 | NP_009169.3 | 3 | 338 | ||
GenBank mRNA | AF072864.1 | - | 3 | 260 | ||
AK023879.1 | - | 3 | 256 | |||
AK301532.1 | - | 3 | 338 | |||
AK316589.1 | - | 3 | 318 | |||
AL136648.1 | - | 3 | 309 | |||
BC001147.1 | - | 3 | 239 | 3533572 | ||
CR533471.1 | - | 3 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
