siRNA Details
Gene Symbol: Ep300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765878.1 | XP_008764100.1 | 1, 2 | 1061 | ||
GenBank mRNA | AB066220.1 | - | 1, 2 | 91 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177821, AK042627, BC150681, BC144976.
Gene Ontology Categories:
