siRNA Details
Gene Symbol: RGD1562392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750614.3 | XP_003750662.1 | 3, 4 | 436 | ||
XM_008766791.1 | XP_008765013.1 | 3, 4 | 415 |
Gene Symbol: LOC100910057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244225.2 | XP_006244287.1 | 3, 4 | 455 | ||
XM_006244227.2 | XP_006244289.1 | 4, 5 | 492 | |||
XM_006244228.2 | XP_006244290.1 | 4, 5 | 797 | |||
XM_006244229.2 | XP_006244291.1 | 4, 5 | 487 | |||
XM_006244230.2 | XP_006244292.1 | 4, 5 | 792 | |||
XM_006244231.2 | XP_006244293.1 | 3, 4 | 433 | |||
XM_006244232.2 | XP_006244294.1 | 3, 4 | 413 | |||
XM_006244233.2 | XP_006244295.1 | 3, 4 | 718 | |||
XM_006244236.2 | XP_006244298.1 | 1, 2 | 151 | |||
XM_008766781.1 | XP_008765003.1 | 4, 5 | 492 | |||
XM_008766782.1 | XP_008765004.1 | 4, 5 | 475 | |||
XM_008766783.1 | XP_008765005.1 | 4, 5 | 470 | |||
XM_008766784.1 | XP_008765006.1 | 4, 5 | 448 | |||
XM_008766785.1 | XP_008765007.1 | 4, 5 | 492 | |||
XM_008766786.1 | XP_008765008.1 | 4, 5 | 492 |
Gene Symbol: Sult1c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013177.2 | NP_001013195.2 | 428 | |||
GenBank mRNA | AJ238392.1 | - | 369 | |||
BC081691.1 | - | 325 | 7110136 | |||
BC092564.1 | - | 428 | 7372924 | |||
FQ209913.1 | - | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
