siRNA Details
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133547.4 | NP_598231.3 | 7 | 742 | ||
XM_006244242.2 | XP_006244304.1 | 6 | 718 | |||
XM_006244243.2 | XP_006244305.1 | 6 | 710 | |||
XM_006244245.2 | XP_006244307.1 | 6 | 705 | |||
XM_006244246.2 | XP_006244308.1 | 6 | 693 | |||
XM_006244248.2 | XP_006244310.1 | 7 | 835 | |||
XM_006244249.2 | XP_006244311.1 | 5 | 644 | |||
XM_006244250.2 | XP_006244312.1 | 5 | 753 | |||
XM_008766738.1 | XP_008764960.1 | 6 | 723 | |||
XM_008766739.1 | XP_008764961.1 | 6 | 686 | |||
XM_008766740.1 | XP_008764962.1 | 6 | 716 | |||
XM_008766741.1 | XP_008764963.1 | 6 | 711 | |||
XM_008766742.1 | XP_008764964.1 | 6 | 698 | |||
XM_008766743.1 | XP_008764965.1 | 6 | 741 | |||
XM_008766744.1 | XP_008764966.1 | 5 | 631 | |||
XM_008766745.1 | XP_008764967.1 | 5 | 619 | |||
GenBank mRNA | AJ238391.1 | - | 4 | 524 | ||
BC103636.1 | - | 7 | 742 | 7106590 |
Gene Symbol: LOC100910057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244225.2 | XP_006244287.1 | 5 | 670 | ||
XM_006244227.2 | XP_006244289.1 | 6 | 707 | |||
XM_006244228.2 | XP_006244290.1 | 6 | 1012 | |||
XM_006244229.2 | XP_006244291.1 | 6 | 702 | |||
XM_006244230.2 | XP_006244292.1 | 6 | 1007 | |||
XM_006244231.2 | XP_006244293.1 | 5 | 648 | |||
XM_006244232.2 | XP_006244294.1 | 5 | 628 | |||
XM_006244233.2 | XP_006244295.1 | 5 | 933 | |||
XM_006244236.2 | XP_006244298.1 | 3 | 366 | |||
XM_008766781.1 | XP_008765003.1 | 6 | 707 | |||
XM_008766782.1 | XP_008765004.1 | 6 | 690 | |||
XM_008766783.1 | XP_008765005.1 | 6 | 685 | |||
XM_008766784.1 | XP_008765006.1 | 6 | 663 | |||
XM_008766785.1 | XP_008765007.1 | 6 | 707 | |||
XM_008766786.1 | XP_008765008.1 | 6 | 707 |
Gene Symbol: LOC100910526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244254.2 | XP_006244316.1 | 553 |
Gene Symbol: RGD1559960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244256.2 | XP_006244318.2 | 2 | 327 | ||
XM_008766792.1 | XP_008765014.1 | 3 | 474 |
Gene Symbol: RGD1562392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750614.3 | XP_003750662.1 | 5 | 651 | ||
XM_008766791.1 | XP_008765013.1 | 5 | 630 |
Gene Symbol: Sult1c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013177.2 | NP_001013195.2 | 643 | |||
GenBank mRNA | AJ238392.1 | - | 584 | |||
BC081691.1 | - | 540 | 7110136 | |||
BC092564.1 | - | 643 | 7372924 | |||
FQ209913.1 | - | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
