siRNA Details
Gene Symbol: Zfp422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012745.1 | NP_001012763.1 | 2 | 507 | ||
XM_006237187.2 | XP_006237249.1 | 2 | 499 | |||
XM_006237189.2 | XP_006237251.1 | 2 | 327 | |||
XM_006237190.2 | XP_006237252.1 | 2 | 358 | |||
XM_008763249.1 | XP_008761471.1 | 2 | 435 | |||
GenBank mRNA | AF281635.1 | - | 1 | 211 | ||
AY724498.1 | - | 2 | 329 | |||
BC090354.1 | - | 2 | 507 | 7319307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
