siRNA Details
Gene Symbol: Khsrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133602.1 | NP_598286.1 | 9, 10 | 875 | ||
XM_006244292.2 | XP_006244354.1 | 9, 10 | 1013 | |||
GenBank mRNA | AF308818.1 | - | 9, 10 | 875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
