siRNA Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000844.3 | NP_000835.1 | 10 | 3969 | ||
NM_181874.2 | NP_870989.1 | 11 | 4061 | |||
XM_017006272.1 | XP_016861761.1 | 11 | 3527 | |||
XM_017006273.1 | XP_016861762.1 | 10 | 3435 | |||
GenBank mRNA | AK123326.1 | - | 1 | 2780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
