siRNA Details
Gene Symbol: Dnajc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138856.2 | NP_620211.2 | 3 | 286 | ||
XM_008760746.1 | XP_008758968.1 | 7 | 1162 | |||
GenBank mRNA | AF492384.1 | - | 2 | 209 | ||
BC086587.1 | - | 3 | 286 | |||
BC158554.1 | - | 3 | 505 | 7380100 |

More Information
Other Species Targeted

Gene Ontology Categories:
