siRNA Details
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047893.3 | NP_001041358.2 | 10 | 6474 | ||
XM_006256816.2 | XP_006256878.1 | 10 | 6556 | |||
XM_006256817.2 | XP_006256879.1 | 10 | 6553 | |||
XM_008773126.1 | XP_008771348.1 | 9 | 6439 | |||
XM_008773127.1 | XP_008771349.1 | 6 | 4031 | |||
XM_008773128.1 | XP_008771350.1 | 6 | 5699 | |||
XM_008773129.1 | XP_008771351.1 | 6 | 5681 | |||
XM_008773130.1 | XP_008771352.1 | 6 | 5684 | |||
XM_008773131.1 | XP_008771353.1 | 6 | 4013 | |||
XM_008773132.1 | XP_008771354.1 | 6 | 4016 | |||
GenBank mRNA | AF348365.1 | - | 1 | 446 | ||
BC085701.1 | - | 1 | 782 | |||
FQ211812.1 | - | 6 | 3581 |

More Information
Other Species Targeted

Gene Ontology Categories:
