siRNA Details
Gene Symbol: Hspb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138887.1 | NP_620242.1 | 2 | 219 | ||
GenBank mRNA | D29960.1 | - | 2 | 219 | ||
FQ214730.1 | - | 2 | 244 | |||
FQ215336.1 | - | 2 | 248 | |||
FQ217608.1 | - | 2 | 248 | |||
FQ224974.1 | - | 2 | 248 |
