siRNA Details
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138913.1 | NP_620268.1 | 4 | 789 | ||
XM_006249382.2 | XP_006249444.1 | 4 | 833 | |||
GenBank mRNA | AY221507.1 | - | 4 | 805 | ||
BC086979.1 | - | 4 | 776 | 7191865 | ||
FQ231421.1 | - | 4 | 818 | |||
FQ232784.1 | - | 4 | 821 | |||
Z18877.1 | - | 4 | 807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
