siRNA Details
Gene Symbol: Sycn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139086.1 | NP_620786.1 | 1 | 342 | ||
GenBank mRNA | AF008197.1 | - | 1 | 342 | ||
AF012887.1 | - | 322 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sycn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139086.1 | NP_620786.1 | 1 | 342 | ||
GenBank mRNA | AF008197.1 | - | 1 | 342 | ||
AF012887.1 | - | 322 |