siRNA Details
Gene Symbol: Tpcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139332.3 | NP_647548.2 | 28 | 2754 | ||
GenBank mRNA | AB018253.2 | - | 27 | 2757 | ||
BC062072.1 | - | 27 | 2754 | 5599545 | ||
CK473123.1 | - | 3 | 332 |
