siRNA Details
Gene Symbol: Slc34a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139338.2 | NP_647554.1 | 10, 11 | 1176 | ||
XM_006233558.2 | XP_006233620.1 | 11, 12 | 1389 | |||
XM_006233559.2 | XP_006233621.1 | 11, 12 | 1505 | |||
XM_006233560.2 | XP_006233622.1 | 11, 12 | 1366 | |||
XM_006233561.2 | XP_006233623.1 | 10, 11 | 1279 | |||
XM_006233562.2 | XP_006233624.1 | 10, 11 | 1395 | |||
XM_006233564.2 | XP_006233626.1 | 10, 11 | 1259 | |||
XM_006233565.2 | XP_006233627.1 | 11, 12 | 1365 | |||
XM_006233566.2 | XP_006233628.1 | 10, 11 | 1371 | |||
XM_006233567.2 | XP_006233629.1 | 10, 11 | 1247 | |||
XM_008761577.1 | XP_008759799.1 | 11, 12 | 1454 | |||
GenBank mRNA | AB077042.1 | - | 10, 11 | 1181 |
