siRNA Details
Gene Symbol: Oas1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009680.1 | NP_001009680.1 | 5 | 961 | ||
GenBank mRNA | AY196697.1 | - | 5 | 961 | ||
BC160819.1 | - | 5 | 935 | 7382468 |
Gene Symbol: LOC100910735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751197.2 | XP_003751245.1 | 981 |
Gene Symbol: Oas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144752.2 | NP_653353.2 | 5 | 955 | ||
GenBank mRNA | AF068268.1 | - | 967 | |||
BC091121.1 | - | 5 | 955 | 7383389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
