siRNA Details
Gene Symbol: Slc15a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144758.1 | NP_653359.1 | 1 | 168 | ||
GenBank mRNA | AB000280.1 | - | 1 | 168 | ||
BC087709.1 | - | 1 | 167 | 7315369 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc15a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144758.1 | NP_653359.1 | 1 | 168 | ||
GenBank mRNA | AB000280.1 | - | 1 | 168 | ||
BC087709.1 | - | 1 | 167 | 7315369 |