siRNA Details
Gene Symbol: Cep104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145082.2 | NP_659550.2 | 11 | 1657 | ||
GenBank mRNA | U53513.1 | - | 2 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
