siRNA Details
Gene Symbol: AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004065.4 | NP_001004065.2 | 2 | 311 | ||
NM_001136562.2 | NP_001130034.1 | 2 | 149 | |||
NM_001198656.1 | NP_001185585.1 | 2 | 311 | |||
GenBank mRNA | AK057098.1 | - | 2 | 213 | ||
AK300427.1 | - | 2 | 124 | |||
AK304311.1 | - | 2 | 193 | |||
AK308388.1 | - | 2 | 273 | |||
BC146863.1 | - | 2 | 311 | 9056791 | ||
BC171800.1 | - | 2 | 311 | 9054454 | ||
BE674805.1 | - | 2 | 110 | |||
DC422367.1 | - | 2 | 149 |
Gene Symbol: PALM2-AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007203.4 | NP_009134.1 | 8 | 827 | ||
NM_147150.2 | NP_671492.1 | 8 | 827 | |||
GenBank mRNA | AB023137.1 | - | 8 | 827 | ||
AJ303079.1 | - | 8 | 815 | |||
BC140818.1 | - | 8 | 668 | 9021686 | ||
BX649126.1 | - | 6 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
