siRNA Details
Gene Symbol: Slc4a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152938.1 | NP_690921.1 | 11, 12 | 1673 | ||
XM_006254530.2 | XP_006254592.1 | 11, 12 | 1817 | |||
XM_006254533.2 | XP_006254595.1 | 11, 12 | 1818 | |||
XM_008772020.1 | XP_008770242.1 | 11, 12 | 1817 | |||
XM_008772021.1 | XP_008770243.1 | 11, 12 | 1818 | |||
XM_008772022.1 | XP_008770244.1 | 11, 12 | 1817 | |||
XM_008772023.1 | XP_008770245.1 | 10, 11 | 1603 | |||
GenBank mRNA | AB024339.1 | - | 11, 12 | 1673 |
