siRNA Details
Gene Symbol: Ugt2b17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153314.2 | NP_695226.2 | 1634 | |||
GenBank mRNA | M31109.1 | - | 1634 | |||
Y00156.1 | - | 6 | 1678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
