siRNA Details
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153622.2 | NP_705886.1 | 8 | 1021 | ||
XM_006253941.2 | XP_006254003.1 | 11 | 1674 | |||
XM_006253942.2 | XP_006254004.1 | 11 | 1676 | |||
XM_006253944.2 | XP_006254006.1 | 11 | 1663 | |||
XM_006253946.2 | XP_006254008.1 | 10 | 1550 | |||
XM_006253947.2 | XP_006254009.1 | 11 | 1680 | |||
XM_006253948.2 | XP_006254010.1 | 6 | 1551 | |||
XM_006253949.2 | XP_006254011.1 | 9 | 1433 | |||
XM_006253950.2 | XP_006254012.1 | 10 | 1443 | |||
XM_006253951.2 | XP_006254013.1 | 9 | 1317 | |||
XM_008771623.1 | XP_008769845.1 | 12 | 1751 | |||
XM_008771624.1 | XP_008769846.1 | 12 | 1739 | |||
XM_008771625.1 | XP_008769847.1 | 10 | 1509 | |||
GenBank mRNA | AB025223.1 | - | 8 | 1045 | ||
AB025224.1 | - | 7 | 958 | |||
BC078710.1 | - | 8 | 939 | 7110091 |
