siRNA Details
Gene Symbol: Slc12a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153625.1 | NP_705889.1 | 9 | 1520 | ||
XM_006248422.2 | XP_006248484.1 | 10 | 1700 | |||
XM_006248423.2 | XP_006248485.1 | 10 | 1665 | |||
XM_006248424.2 | XP_006248486.1 | 10 | 1949 | |||
XM_006248425.2 | XP_006248487.1 | 10 | 1538 | |||
XM_006248426.2 | XP_006248488.1 | 9 | 1586 | |||
XM_006248427.2 | XP_006248489.1 | 9 | 1553 | |||
XM_006248429.2 | XP_006248491.1 | 10 | 1700 | |||
XM_008768704.1 | XP_008766926.1 | 10 | 1622 | |||
XM_008768706.1 | XP_008766928.1 | 10 | 1622 | |||
GenBank mRNA | AB024430.1 | - | 9 | 1520 | ||
AB024431.1 | - | 9 | 1442 |
