siRNA Details
Gene Symbol: HTR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000870.6 | NP_000861.1 | 7 | 3282 | ||
NM_001040173.2 | NP_001035263.1 | 7 | 2936 | |||
NM_001286410.1 | NP_001273339.1 | 6 | 1152 | |||
NR_104445.1 | 8 | 3529 | ||||
GenBank mRNA | AJ131724.1 | - | 7 | 2908 | ||
AM712912.1 | - | 6 | 1105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
