siRNA Details
Gene Symbol: Ly49s3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153726.1 | NP_714948.1 | 9 | 1000 | ||
GenBank mRNA | AY102036.1 | - | 9 | 1000 | ||
AY747628.1 | - | 10 | 1051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
