siRNA Details
Gene Symbol: Serpinb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153733.2 | NP_714955.2 | 3 | 255 | ||
GenBank mRNA | AY075037.1 | - | 3 | 271 | ||
BC061735.1 | - | 3 | 255 | 5598648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
