siRNA Details
Gene Symbol: Cdk5rap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173134.2 | NP_775157.2 | 34, 35 | 5393 | ||
XM_006238273.2 | XP_006238335.1 | 35, 36 | 5878 | |||
XM_006238274.2 | XP_006238336.1 | 35, 36 | 5872 | |||
XM_006238276.2 | XP_006238338.1 | 34, 35 | 5780 | |||
XM_006238277.2 | XP_006238339.1 | 34, 35 | 5774 | |||
XM_006238278.2 | XP_006238340.1 | 34, 35 | 5627 | |||
XM_006238279.2 | XP_006238341.1 | 34, 35 | 5620 | |||
XM_006238280.2 | XP_006238342.1 | 33, 34 | 5549 | |||
XM_008763760.1 | XP_008761982.1 | 35, 36 | 5878 | |||
XM_008763761.1 | XP_008761983.1 | 34, 35 | 5747 | |||
GenBank mRNA | AF177478.1 | - | 3, 4 | 431 | ||
JX524852.1 | - | 35, 36 | 5486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
