siRNA Details
Gene Symbol: Lama3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772008.1 | XP_008770230.1 | 39 | 5983 | ||
GenBank mRNA | U61261.1 | - | 2 | 309 |

More Information
Other Species Targeted

Gene Ontology Categories:
