siRNA Details
Gene Symbol: COPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007263.3 | NP_009194.2 | 7 | 647 | ||
NM_199442.1 | NP_955474.1 | 6 | 494 | |||
XM_011527655.1 | XP_011525957.1 | 8 | 790 | |||
GenBank mRNA | AF092438.1 | - | 7 | 642 | ||
AJ131182.1 | - | 7 | 647 | |||
AJ249366.1 | - | 7 | 620 | |||
AK222584.1 | - | 7 | 644 | |||
AK315281.1 | - | 7 | 644 | |||
AL136928.1 | - | 7 | 624 | |||
BC003155.1 | - | 7 | 619 | 2906127 | ||
BC007250.1 | - | 7 | 624 | 2989183 | ||
BC017285.2 | - | 7 | 610 | 4891380 | ||
BI520280.1 | - | 7 | 660 | |||
CR456886.1 | - | 7 | 605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
