siRNA Details
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173332.2 | NP_775454.2 | 2 | 181 | ||
XM_006237141.2 | XP_006237203.1 | 3 | 704 | |||
XM_006237142.2 | XP_006237204.1 | 2 | 1760 | |||
XM_006237143.2 | XP_006237205.1 | 2 | 558 | |||
XM_006237145.2 | XP_006237207.1 | 3 | 1021 | |||
XM_008763224.1 | XP_008761446.1 | 3 | 1022 | |||
XM_008763225.1 | XP_008761447.1 | 4 | 775 | |||
XM_008763226.1 | XP_008761448.1 | 2 | 875 | |||
GenBank mRNA | AF315467.1 | - | 2 | 495 | ||
BC078692.1 | - | 2 | 273 | |||
BC162051.1 | - | 2 | 181 | 7112072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
