siRNA Details
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173332.2 | NP_775454.2 | 4 | 455 | ||
XM_006237141.2 | XP_006237203.1 | 5 | 978 | |||
XM_006237142.2 | XP_006237204.1 | 4 | 2034 | |||
XM_006237143.2 | XP_006237205.1 | 4 | 832 | |||
XM_006237145.2 | XP_006237207.1 | 5 | 1295 | |||
XM_006237146.1 | XP_006237208.1 | 2 | 136 | |||
XM_008763224.1 | XP_008761446.1 | 5 | 1296 | |||
XM_008763225.1 | XP_008761447.1 | 6 | 1049 | |||
XM_008763226.1 | XP_008761448.1 | 4 | 1149 | |||
XM_008763227.1 | XP_008761449.1 | 2 | 263 | |||
GenBank mRNA | AF315467.1 | - | 4 | 769 | ||
BC078692.1 | - | 4 | 547 | |||
BC162051.1 | - | 4 | 455 | 7112072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
