siRNA Details
Gene Symbol: Hs3st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181370.2 | NP_852035.1 | 2 | 1647 | ||
GenBank mRNA | AY240873.1 | - | 2 | 1647 | ||
BC128721.1 | - | 2 | 1650 | 8364509 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hs3st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181370.2 | NP_852035.1 | 2 | 1647 | ||
GenBank mRNA | AY240873.1 | - | 2 | 1647 | ||
BC128721.1 | - | 2 | 1650 | 8364509 |