siRNA Details
Gene Symbol: Rtn4rl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181377.3 | NP_852042.1 | 2 | 838 | ||
GenBank mRNA | AF532861.1 | - | 2 | 593 | ||
AY311478.1 | - | 3 | 701 | |||
BC129084.1 | - | 2 | 838 | 8363864 |
