siRNA Details
Gene Symbol: ABL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005157.5 | NP_005148.2 | 3 | 682 | ||
NM_007313.2 | NP_009297.2 | 3 | 986 | |||
GenBank mRNA | AB209456.1 | - | 3 | 692 | ||
AB209642.1 | - | 3 | 915 | |||
BC117451.1 | - | 3 | 612 | 40126002 | ||
M14752.1 | - | 3 | 854 | |||
X16416.1 | - | 3 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
