siRNA Details
Gene Symbol: Tsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021854.1 | NP_068626.1 | 2, 3 | 221 | ||
XM_006233843.2 | XP_006233905.1 | 4, 5 | 675 | |||
XM_006233844.2 | XP_006233906.1 | 4, 5 | 646 | |||
XM_006233845.2 | XP_006233907.1 | 4, 5 | 497 | |||
XM_006233846.2 | XP_006233908.1 | 4, 5 | 2762 | |||
XM_006233847.2 | XP_006233909.1 | 4, 5 | 983 | |||
XM_006233849.2 | XP_006233911.1 | 4, 5 | 675 | |||
GenBank mRNA | AB011821.1 | - | 2, 3 | 221 |
