siRNA Details
Gene Symbol: Syt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022191.1 | NP_071527.1 | 3 | 1176 | ||
GenBank mRNA | BC099185.1 | - | 832 | 7383303 | ||
U20105.1 | - | 3 | 1176 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001276681, NM_018800, NM_001276680, NM_001276679, NM_001276677, and others.
Gene Ontology Categories:
