siRNA Details
Gene Symbol: HSP90AB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271969.1 | NP_001258898.1 | 3 | 499 | ||
NM_001271970.1 | NP_001258899.1 | 3 | 427 | |||
NM_001271972.1 | NP_001258901.1 | 3 | 486 | |||
NM_007355.3 | NP_031381.2 | 3 | 486 | |||
GenBank mRNA | AK294643.1 | - | 3 | 343 | ||
AK297367.1 | - | 3 | 343 | |||
AK312255.1 | - | 3 | 347 | |||
AL136543.1 | - | 3 | 322 | |||
AY359878.1 | - | 3 | 360 | |||
BC004928.1 | - | 3 | 332 | 3621040 | ||
BC007327.2 | - | 3 | 324 | |||
BC009206.2 | - | 1 | 26 | |||
BC012807.2 | - | 3 | 332 | 3530042 | ||
BC014485.1 | - | 3 | 327 | 4870198 | ||
BC016753.2 | - | 3 | 345 | 2987963 | ||
BC068474.1 | - | 3 | 350 | 4824686 | ||
BI252490.1 | - | 1 | 333 | |||
BX420114.2 | - | 3 | 387 | |||
BX649079.1 | - | 3 | 339 | |||
CX761569.1 | - | 3 | 376 | |||
DC340766.1 | - | 3 | 419 | |||
DC341882.1 | - | 3 | 435 | |||
DC350708.1 | - | 3 | 486 | |||
DC365600.1 | - | 3 | 421 | |||
M16660.1 | - | 3 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
