siRNA Details
Gene Symbol: Arl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022385.3 | NP_071780.1 | 5 | 456 | ||
GenBank mRNA | BC061553.1 | - | 539 | 6889396 | ||
FQ226660.1 | - | 5 | 512 | |||
U12402.1 | - | 5 | 388 | |||
X76920.1 | - | 5 | 456 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025859, AK214089, AK010732, AK088330, AK207710, and others.
Gene Ontology Categories:
