siRNA Details
Gene Symbol: Pttg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022391.2 | NP_071786.1 | 2 | 534 | ||
XM_006246142.1 | XP_006246204.1 | 3 | 345 | |||
XM_006246143.2 | XP_006246205.1 | 3 | 542 | |||
XM_006246144.1 | XP_006246206.1 | 3 | 639 | |||
XM_006246145.2 | XP_006246207.1 | 3 | 345 | |||
XM_008767643.1 | XP_008765865.1 | 3 | 294 | |||
GenBank mRNA | BC158551.1 | - | 3 | 644 | 9032805 | |
U73030.1 | - | 2 | 546 |
