siRNA Details
Gene Symbol: OPRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000914.4 | NP_000905.3 | 4 | 2110 | ||
NM_001145279.3 | NP_001138751.1 | 6 | 2469 | |||
NM_001145280.3 | NP_001138752.1 | 4 | 1900 | |||
NM_001145281.2 | NP_001138753.1 | 4 | 1441 | |||
NM_001145287.2 | NP_001138759.1 | 4 | 2675 | |||
NM_001285522.1 | NP_001272451.1 | 2 | 1236 | |||
NM_001285524.1 | NP_001272453.1 | 5 | 2005 | |||
NM_001285526.1 | NP_001272455.1 | 4 | 2656 | |||
NR_104348.1 | 5 | 1910 | ||||
NR_104350.1 | 3 | 1448 | ||||
XM_017010906.1 | XP_016866395.1 | 4 | 3585 | |||
GenBank mRNA | L25119.1 | - | 4 | 1880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
