siRNA Details
Gene Symbol: Mpeg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022617.1 | NP_072139.1 | 1 | 242 | ||
XM_006231115.2 | XP_006231177.1 | 2 | 319 | |||
XM_008760261.1 | XP_008758483.1 | 2 | 411 | |||
GenBank mRNA | AF156540.1 | - | 1 | 242 |
