siRNA Details
Gene Symbol: Gabarapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022706.2 | NP_073197.1 | 4 | 405 | ||
GenBank mRNA | AB003515.1 | - | 4 | 443 | ||
BC058145.1 | - | 4 | 408 | 6921109 | ||
BC088139.1 | - | 4 | 405 | 7323806 | ||
FQ216852.1 | - | 4 | 419 | |||
FQ226397.1 | - | 4 | 429 | |||
FQ231643.1 | - | 4 | 398 | |||
FQ233551.1 | - | 4 | 440 | |||
FQ234861.1 | - | 4 | 398 |
Gene Symbol: LOC100359937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729947.4 | XP_002729993.3 | 1 | 394 |
Gene Symbol: LOC103692360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763731.1 | XP_008761953.1 | 5 | 775 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026693, BC081436, AK049819, AK134873, AK002596, and others.
Gene Ontology Categories:
