siRNA Details
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022947.1 | NP_075236.1 | 1 | 155 | ||
XM_008759796.1 | XP_008758018.1 | 1 | 419 | |||
GenBank mRNA | AB027570.1 | - | 1 | 155 | ||
BC070917.1 | - | 1 | 118 | 7105142 |
