siRNA Details
Gene Symbol: Rhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023022.2 | NP_075411.2 | 1 | 222 | ||
GenBank mRNA | AB015194.1 | - | 1 | 195 | ||
AF531097.1 | - | 1 | 222 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023022.2 | NP_075411.2 | 1 | 222 | ||
GenBank mRNA | AB015194.1 | - | 1 | 195 | ||
AF531097.1 | - | 1 | 222 |