siRNA Details
Gene Symbol: Cd200r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023953.1 | NP_076443.1 | 3 | 292 | ||
XM_006248347.2 | XP_006248409.1 | 3 | 605 | |||
XM_008768743.1 | XP_008766965.1 | 3 | 592 | |||
GenBank mRNA | AY583211.1 | - | 3 | 292 |
